missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terephthalaldehyde, 98+%

$37.72 - $81.73

Chemical Identifiers

CAS 623-27-8
Molecular Formula C8H6O2
Formula Weight 134.14
MDL Number MFCD00006949
Beilstein 385863
Synonym Benzene-1,4-dicarboxaldehyde; Terephthaldicarboxaldehyde
Chemical Name or Material Terephthalaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1493014 SDS Alfa Aesar™
A1493014*
25g Each for $37.72
AAA1493022 SDS Alfa Aesar™
A1493022
100g Each for $81.73
AAA1493036 SDS Alfa Aesar™
A1493036
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Terephthalaldehyde is used as an intermediate for the preparation of dyes and fluorescent whitening agents. It is used in the synthesis of polyimines by reacting with aliphatic diamines. It is used as a precursor for the preparation of paramagnetic microporous polymeric organic frameworks (POFs) through copolymerization with pyrrole, indole, and carbazole.

Solubility
Soluble in hot water, alcohol and ether.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
Description & Specifications

Chemical Identifiers

623-27-8
134.14
385863
Terephthalaldehyde
C8H6O2
MFCD00006949
Benzene-1,4-dicarboxaldehyde; Terephthaldicarboxaldehyde

Specifications

623-27-8
134.14
385863
245°C
76°C (168°F)
Terephthalaldehyde
C8H6O2
MFCD00006949
Benzene-1,4-dicarboxaldehyde; Terephthaldicarboxaldehyde
114° to 117°C
25g
≥98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WZ0430000

TSCA : TSCA