missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium sputtering target, 50.8mm dia x 3.18mm thick

$2,541.35 - $2,541.35

Chemical Identifiers

CAS 7440-27-9
MDL Number MFCD00011256
Chemical Name or Material Terbium sputtering target, 50.8mm dia. x 3.18mm thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45738KS SDS Alfa Aesar™
45738KS
1 ea. Each for $2,541.35
Specifications Description & Specifications

Chemical Identifiers

7440-27-9
MFCD00011256
Terbium sputtering target, 50.8mm dia. x 3.18mm thick

Specifications

1 ea.
7440-27-9
MFCD00011256
Terbium sputtering target, 50.8mm dia. x 3.18mm thick

Safety and Handling

TSCA : TSCA