missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Terbium Rod, 12.7mm (0.50 in.) dia., 99.9% (Metals basis), Alfa Aesar™

Form: Approx. 1.04g/mm; Cast rod, diameter is approximate

Manufacturer:  Thermo Fisher Scientific 44049DH

 View more versions of this product

Catalog No. AA44049DH



Specifications

Specifications

Terbium
99.9% (Metals basis)
MFCD00011256
acmc-20ajly
[Tb]
158.925
CHEBI:33376
Silver to Gray
7440-27-9
Tb
Silver Gray rod
GZCRRIHWUXGPOV-UHFFFAOYSA-N
terbium
23958
≈1.04g/mm,Cast Rod,diameter is approximate
25mm

Chemical Identifiers

7440-27-9
158.925
GZCRRIHWUXGPOV-UHFFFAOYSA-N
23958
terbium
Tb
MFCD00011256
acmc-20ajly
CHEBI:33376
[Tb]
Safety and Handling

Safety and Handling

EINECSNumber : 231-137-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents