missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01459609

 View more versions of this product

Catalog No. AA1459609


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Terbium(III) sulfate octahydrate
13842-67-6
10 g
MFCD00149872
99.9%
360°C (in -8H2O)
Crystalline
(REO)
750.19 (606.05 Anhydrous)
Reacton™

Chemical Identifiers

13842-67-6
MFCD00149872
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-213-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only