missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium(III) sulfate octahydrate, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 4447918

 View more versions of this product

Catalog No. AA4447918



Specifications

Specifications

Terbium (III) sulfate octahydrate
(metals basis)
MFCD00149872
Crystalline
360°C (in -8H2O)
13842-67-6
Tb2(SO4)3·8H2O
750.19 (606.05 Anhydrous)
99.995%
50g

Chemical Identifiers

13842-67-6
Tb2(SO4)3·8H2O
MFCD00149872
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-213-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents