missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Terbium(III) nitrate hydrate, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 7410306

 View more versions of this product

Catalog No. AA7410306



Specifications

Specifications

Terbium(III) nitrate hydrate
99.9% (REO)
MFCD00011257
UNSPFWLYDUXJHT-UHFFFAOYSA-N
terbium(3+);trinitrate;hydrate
91886285
Wet Crystalline Aggregates, Packaged Under Argon
94219-56-4
H2N3O10Tb
terbium 3+ hydrate trinitrate, 3no3.tb.h2o
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Tb+3]
362.952
344.94 (Anhydrous)
5g

Chemical Identifiers

94219-56-4
362.952
UNSPFWLYDUXJHT-UHFFFAOYSA-N
91886285
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Tb+3]
H2N3O10Tb
MFCD00011257
terbium 3+ hydrate trinitrate, 3no3.tb.h2o
terbium(3+);trinitrate;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : 233-138-7

SDS Documents