missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Terbium(III) nitrate hexahydrate, 99.9%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC295790100

 View more versions of this product

Catalog No. AC295790100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

453.02
Granular Powder
White
99.9%
Tb(NO3)3·6H2O
MFCD00149868
13451-19-9
Trace Metal Basis
10g
0.1% max.
Terbium(III) nitrate hexahydrate
N3O9Tb·6H2O
99.99+%

Chemical Identifiers

13451-19-9
N3O9Tb·6H2O
453.02
Terbium(III) nitrate hexahydrate, 99.9%
99.99+%
Tb(NO3)3·6H2O
MFCD00149868
Trace Metal Basis

Safety and Handling

SDS & Safety Documents

Danger

RTECSNumber : WY9625000