missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium(III,IV) oxide, REacton™, 99.998% (REO)

Manufacturer:  Alfa Aesar™ 1091703

 View more versions of this product

Catalog No. AA1091703



Specifications

Specifications

Terbium(III,IV) oxide
(REO)
14,9157
Powder
Reacton™
2340°C
12037-01-3
MFCD00083151
747.69
99.998%
Odorless
1g

Chemical Identifiers

12037-01-3
MFCD00083151
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents