missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium(III) chloride hexahydrate, REacton™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4447206

 View more versions of this product

Catalog No. AA4447206



Specifications

Specifications

Terbium (III) chloride hexahydrate
(metals basis)
MFCD00149870
Very soluble in water.
Crystalline
Reacton™
5g
13798-24-8
TbCl3·6H2O
14,9157
373.38 (265.29 Anhydrous)
99.999%
Odorless

Chemical Identifiers

13798-24-8
TbCl3·6H2O
MFCD00149870
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 233-132-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents