missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Terbium(III) chloride hexahydrate, 99.9%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 199610250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisSpecifications
Terbium(III) chloride hexahydrate | |
25g | |
Cl3Tb·6H2O | |
MFCD00149870 | |
Solubility in water: soluble. Other solubilities: readily soluble in alcohol | |
373.38 | |
Trace Metal Basis |
13798-24-8 | |
99.90% | |
TbCl3·6H2O | |
14, 9157 | |
373.38 | |
99.9% | |
Glass bottle |
RUO â Research Use Only