Learn More
Terbium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01120918
Description
Terbium(III) chloride hexahydrate is a water soluble source used for the preparation of metallic terbium, which is used in electronic devices and an additive for special alloy namely Terfenol-D. It is used in the preparation of complexes, which are used in the spectrofluorimetric determination of various chemicals in pharmaceutical formulations. Further, it finds use in the preparation of terbium nanotubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Terbium(III) chloride hexahydrate | |
Crystalline Aggregates | |
99.9% (REO) | |
MFCD00149870 | |
ULJUVCOAZNLCJZ-UHFFFAOYSA-K | |
terbium(3+) hexahydrate trichloride | |
373.38 (265.29 Anhydrous) |
13798-24-8 | |
50 g | |
Cl3H12O6Tb | |
Very soluble in water. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3] | |
373.37 | |
Hygroscopic |
Chemical Identifiers
13798-24-8 | |
373.37 | |
ULJUVCOAZNLCJZ-UHFFFAOYSA-K | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3] |
Cl3H12O6Tb | |
MFCD00149870 | |
terbium(3+) hexahydrate trichloride |
Safety and Handling
EINECSNumber : 233-132-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only