Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium(III) carbonate hydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Terbium(III) carbonate hydrate
CAS 100587-96-0
Purity Grade Reacton™
Formula Weight 490.91 (Anhydrous)
MDL Number MFCD00149234
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1459909

  SDS
alfa aesar™
1459909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1459918

  SDS
alfa aesar™
1459918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 490.91 (Anhydrous)
Quantity 10g
MDL Number MFCD00149234
Chemical Name or Material Terbium(III) carbonate hydrate
TSCA TSCA
Sensitivity Hygroscopic
CAS 100587-96-0
Purity Grade Reacton™
Physical Form Crystalline
Assay 99.9%
Assay Percent Notes (REO)