missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium(III) carbonate hydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1459918

 View more versions of this product

Catalog No. AA1459918



Specifications

Specifications

Terbium(III) carbonate hydrate
(REO)
MFCD00149234
BIKXNTALUHUKPA-UHFFFAOYSA-H
terbium(3+);tricarbonate;hydrate
57346090
Crystalline
Reacton™
50g
100587-96-0
C3H2O10Tb2
terbium iii carbonate hydrate, acmc-20ajlq, terbium iii carbonate hydrate, reacton, terbium 3+ carbonate-water 2/3/1, carbonic acid,terbium 3+ salt 3:2 , hydrate 9ci
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Tb+3].[Tb+3]
515.89
490.91 (Anhydrous)
99.9%
Hygroscopic

Chemical Identifiers

100587-96-0
515.89
BIKXNTALUHUKPA-UHFFFAOYSA-H
57346090
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Tb+3].[Tb+3]
C3H2O10Tb2
MFCD00149234
terbium iii carbonate hydrate, acmc-20ajlq, terbium iii carbonate hydrate, reacton, terbium 3+ carbonate-water 2/3/1, carbonic acid,terbium 3+ salt 3:2 , hydrate 9ci
terbium(3+);tricarbonate;hydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents