Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium(III) bromide, ultra dry, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Terbium(III) bromide
CAS 14456-47-4
Name Note Ultra dry
Formula Weight 398.65
MDL Number MFCD00049571
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3942503

  SDS
alfa aesar™
3942503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3942506

  SDS
alfa aesar™
3942506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 398.65
Packaging Ampule
TSCA TSCA
Sensitivity Hygroscopic
Chemical Name or Material Terbium(III) bromide
CAS 14456-47-4
Name Note Ultra dry
MDL Number MFCD00049571
Physical Form Powder
Storage Note 1
  • Ampouled under argon
Assay Percent Notes (REO)
Boiling Point 1490°C
Assay 99.99%
Melting Point 827°C
Quantity 1g

  • Ampouled under argon

Safety and Handling

  • Ampouled under argon