Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium(III) bromide hydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Terbium(III) bromide hydrate
Purity Grade Reacton™
Formula Weight 398.65 (Anhydrous)
MDL Number MFCD00049571
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1317309

  SDS
alfa aesar™
1317309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1317318

  SDS
alfa aesar™
1317318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 398.65 (Anhydrous)
Chemical Name or Material Terbium(III) bromide hydrate
TSCA TSCA
Purity Grade Reacton™
Physical Form Crystalline
Assay 99.99%
Assay Percent Notes (REO)
MDL Number MFCD00049571
Melting Point 170°C (dec.)
Quantity 10g