Please login to your online account to display your discounted pricing

Terbium(III) acetate hydrate, 99.9% (REO), Alfa Aesar™, Reacton™

$164.24 - $164.24

Chemical Identifiers

Chemical Name or Material Terbium(III) acetate hydrate
CAS 100587-92-6
Physical Form Crystalline Aggregates
Assay Percent Range 99.9% (REO)
Molecular Formula Tb(OOCCH3)3·xH2O
Formula Weight 336.08 (anhydrous)
MDL Number MFCD00149233
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1460009

  SDS
alfa aesar
1460009
10g Each for $164.24

AA1460018

  SDS
alfa aesar
1460018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Terbium(III) acetate hydrate
CAS 100587-92-6
Physical Form Crystalline Aggregates
Assay Percent Range 99.9% (REO)
Molecular Formula Tb(OOCCH3)3·xH2O
Formula Weight 336.08 (anhydrous)
MDL Number MFCD00149233
Quantity 10g