Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium foil, 0.62mm (0.024 in.) thick, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Terbium foil
CAS 7440-27-9
Name Note 0.62mm (0.024 in.) thick
MDL Number MFCD00011256
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39695FF

 
alfa aesar™
39695FF
25 x 25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39695FI

 
alfa aesar™
39695FI
50 x 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25 x 25mm
Packaging Mylar under Argon
Chemical Name or Material Terbium foil
TSCA TSCA
Name Note 0.62mm (0.024 in.) thick
Physical Form Foil
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011256
CAS 7440-27-9
Storage Note 1
  • Packaged under argon
Weight ≈3.2g/25x25mm

  • Packaged under argon

Safety and Handling

  • Packaged under argon