missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Terbium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 012429FF

 View more versions of this product

Catalog No. AA12429FF


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Terbium foil
7440-27-9
Tb
25 x 25 mm
GZCRRIHWUXGPOV-UHFFFAOYSA-N
terbium
23958
99.9%
≈1.29g/25x25mm
0.25mm (0.01 in.) thick
(REO)
MFCD00011256
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
[Tb]
158.93
CHEBI:33376
Mylar under Argon
Foil

Chemical Identifiers

7440-27-9
158.93
GZCRRIHWUXGPOV-UHFFFAOYSA-N
23958
terbium
Tb
MFCD00011256
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
CHEBI:33376
[Tb]
Safety and Handling

Safety and Handling

EINECSNumber : 231-137-6

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only