missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Terbium foil, 0.25mm (0.01 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12429FI

 View more versions of this product

Catalog No. AA12429FI



Description

Description

  • Packaged under argon
Specifications

Specifications

Terbium foil
50 x 50mm
7440-27-9
(REO)
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
terbium
23958
Foil
≈1.29g/25x25mm
0.25mm (0.01 in.) thick
Mylar under Argon
Tb
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
[Tb]
158.925
CHEBI:33376
99.9%

Chemical Identifiers

7440-27-9
158.925
GZCRRIHWUXGPOV-UHFFFAOYSA-N
23958
terbium
Tb
MFCD00011256
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
CHEBI:33376
[Tb]
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents