missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Terbium Foil, 0.1mm (0.004 in.) thick, (REO), 0.999, Alfa Aesar™

Form: Approx. 0.52g/25 x 25mm

Manufacturer:  Alfa Aesar 12430FI

 View more versions of this product

Catalog No. AA12430FI



Specifications

Specifications

Terbium
99.9% (REO)
MFCD00011256
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
[Tb]
158.925
CHEBI:33376
Silver to Gray
7440-27-9
Tb
Silver Gray foil
GZCRRIHWUXGPOV-UHFFFAOYSA-N
terbium
23958
Foil, 0.1mm (0.004 in.) thick
50 x 50mm

Chemical Identifiers

7440-27-9
158.925
GZCRRIHWUXGPOV-UHFFFAOYSA-N
23958
terbium
Tb
MFCD00011256
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
CHEBI:33376
[Tb]
Safety and Handling

Safety and Handling

H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-137-6

Documents