Please login to your online account to display your discounted pricing

Terbium Foil, 0.025mm (0.001 in.) thick, (REO), 0.999, Alfa Aesar™

Form: Approx. 0.13g/25 x 25mm

Chemical Identifiers

Chemical Name or Material Terbium
CAS 7440-27-9
Physical Form Foil, 0.025mm (0.001 in.) thick
Assay Percent Range 99.9% (REO)
MDL Number MFCD00011256
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12431FF

 
alfa aesar
12431FF
25 x 25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12431FI

 
alfa aesar
12431FI
50 x 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25 x 25mm
Color Silver-gray
DOT Information DOT Class: 4.1, Packaging Group: II
Chemical Name or Material Terbium
EINECS Number 231-137-6
Physical Form Foil, 0.025mm (0.001 in.) thick
Assay Percent Range 99.9% (REO)
Appearance Silver Gray foil
MDL Number MFCD00011256
CAS 7440-27-9
Health Hazard 1 H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.
Safety and Handling

H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.