missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TER-119, PE-eFluor 610, clone: TER-119, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61592180

 View more versions of this product

Catalog No. 501124852



Description

Description

TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.
Specifications

Specifications

TER-119
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Liquid
Ly76
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
104231
Flow Cytometry
TER-119
PE-eFluor® 610
Ly76
TER119; Erythroid cell marker; Ly-76; Ly76
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Documents

For Research Use Only.