missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TER-119 Rat anti-Mouse, PE-Cy5, Clone: TER-119, eBioscience™

Rat Monoclonal Antibody

$196.68 - $496.32

Specifications

Antigen TER-119
Clone TER-119
Host Species Rat
Gene Alias TER119, Erythroid cell marker, Ly-76, Ly76
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122932 Life Technologies
LS15592182
100μg Each for $335.28
5014146 Life Technologies
LS15592183
200μg Each for $496.32
5014144 Life Technologies
LS15592181
50μg Each for $196.68
Specifications

Description

The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin.
Description & Specifications

Specifications

TER-119
Rat
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
TER-119
TER119, Erythroid cell marker, Ly-76, Ly76
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer