Please login to your online account to display your discounted pricing

TER-119 Rat anti-Mouse, FITC, Clone: TER-119, eBioscience™

Rat Monoclonal Antibody

$114.55 - $361.05

Specifications

Antigen TER-119
Clone TER-119
Host Species Rat
Gene Alias TER119, Erythroid cell marker, Ly-76, Ly76
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122151

 
life technologies
LS11592185
500μg Each for $361.05

509916

 
life technologies
LS11592181
50μg Each for $114.55

501129690

 
life technologies
LS11592182
100μg Each for $172.55
Description & Specifications

Specifications

Antigen TER-119
Clone TER-119
Host Species Rat
Gene Alias TER119, Erythroid cell marker, Ly-76, Ly76
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin.