TentaGel™ S-NH2, Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals 354610050
 View more versions of this product
Catalog No. AC354610050
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
Specifications
Specifications
White to Yellow
Powder
Glass bottle
5 g
O-(2-Aminoethyl)polyethylene glycol polymer bound
TentaGel™ S-NH2
0.2 to 0.3mmol/g
90 Micron
MFCD00217899