Please login to your online account to display your discounted pricing

TentaGel™ S-NH2, ACROS Organics™

$320.40 - $320.40

Chemical Identifiers

Chemical Name or Material TentaGel™ S-NH2
MDL Number MFCD00217899
Synonym O-(2-Aminoethyl)polyethylene glycol polymer bound
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC354610050

  SDS
thermo fisher scientific
354610050
Glass bottle 5g Each for $320.40

AC354610250

  SDS
thermo fisher scientific
354610250
Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 5g
Color White to light yellow
Particle Size 90 Micron
Packaging Glass bottle
Loading 0.2 to 0.3mmol/g
Synonym O-(2-Aminoethyl)polyethylene glycol polymer bound
Chemical Name or Material TentaGel™ S-NH2
MDL Number MFCD00217899

For solid support synthesis