TentaGel™ S-NH2, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 354610250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For solid support synthesisSpecifications
TentaGel™ S-NH2 | |
Glass Bottle | |
25g | |
O-(2-Aminoethyl)polyethylene glycol polymer bound |
0.2 to 0.3mmol/g | |
90 Micron | |
MFCD00217899 |
RUO â Research Use Only