TentaGel™ MB-S-Trityl, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 354550010
Description
For solid support synthesisSpecifications
TentaGel™ MB-S-Trityl | |
Glass bottle | |
1g | |
O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound |
0.4 to 0.6mmol/g | |
140 to 170 Micron | |
MFCD01868636 |
RUO â Research Use Only