missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TentaGel™ MB-S-Trityl, ACROS Organics™

Chemical Identifiers

MDL Number MFCD01868636
Synonym O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound
Chemical Name or Material TentaGel™ MB-S-Trityl
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC354550050 SDS Thermo Fisher Scientific
354550050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC354550010 SDS Thermo Fisher Scientific
354550010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

For solid support synthesis
Description & Specifications

Chemical Identifiers

MFCD01868636
O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound
TentaGel™ MB-S-Trityl

Specifications

Powder
White to yellow
MFCD01868636
0.4 to 0.6mmol/g
TentaGel™ MB-S-Trityl
5g
140 to 170 Micron
Glass bottle
O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound