Please login to your online account to display your discounted pricing

TentaGel™ MB-CH(OEt)2, ACROS Organics™

Chemical Identifiers

Chemical Name or Material TentaGel™ MB-CH(OEt)2
MDL Number MFCD01868634
Synonym O-[2-(6, 6-Diethoxyhexanoylamino)ethyl]polyethylene glycol polymer bound
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC354490010

  SDS
thermo fisher scientific
354490010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC354490050

  SDS
thermo fisher scientific
354490050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 1g
Color White to light yellow
Particle Size 140 to 170 Micron
Loading 0.4 to 0.6mmol/g
Packaging Glass bottle
Chemical Name or Material TentaGel™ MB-CH(OEt)2
MDL Number MFCD01868634
Synonym O-[2-(6, 6-Diethoxyhexanoylamino)ethyl]polyethylene glycol polymer bound

For solid support synthesis