Please login to your online account to display your discounted pricing

TentaGel™ HL-S-Trityl, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  354470050

 View more versions of this product

Catalog No. AC354470050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 5g
Color White to light yellow
Particle Size mean particle size: 110 Micron
MDL Number MFCD01868633
Loading 0.4 to 0.6mmol/g
Packaging Glass bottle
Chemical Name or Material TentaGel™ HL-S-Trityl
Synonym O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound

Specifications

Chemical Name or Material TentaGel™ HL-S-Trityl
MDL Number MFCD01868633
Synonym O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound