Please login to your online account to display your discounted pricing

TentaGel™ HL-S-Trityl, ACROS Organics™

Chemical Identifiers

Chemical Name or Material TentaGel™ HL-S-Trityl
MDL Number MFCD01868633
Synonym O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC354470010

  SDS
thermo fisher scientific
354470010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC354470050

  SDS
thermo fisher scientific
354470050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Quantity 1g
Color White to light yellow
Particle Size mean particle size: 110 Micron
Chemical Name or Material TentaGel™ HL-S-Trityl
MDL Number MFCD01868633
Synonym O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound
Loading 0.4 to 0.6mmol/g
Packaging Glass bottle

For solid support synthesis