Please login to your online account to display your discounted pricing

TentaGel™ HL-RAM, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  354460010

 View more versions of this product

Catalog No. AC354460010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 1g
Color White to yellow
Particle Size mean particle size: 75 Micron
Chemical Name or Material TentaGel™ HL-RAM
MDL Number MFCD00217896
Synonym N-FMOC-4'-[poly(oxyethylene)carbamoylmethoxy]-2, 4-dimethoxy-benzhydrylamine polymer bound
Packaging Glass bottle
Loading 0.3 to 0.4mmol/g

Specifications

Chemical Name or Material TentaGel™ HL-RAM
MDL Number MFCD00217896
Synonym N-FMOC-4'-[poly(oxyethylene)carbamoylmethoxy]-2, 4-dimethoxy-benzhydrylamine polymer bound