missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tempshield™ Cryo-Gloves™ Pink Waterproof Shoulder-Length Cryogenic Gloves
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
Supplier: Tempshield™ TSIPSHLWP
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Description
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Pink | |
Fabric | |
Large | |
Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes | |
-320°F | |
24.5 to 27.3 in. |
Shoulder Length | |
Specific | |
Waterproof Cryogenic Gloves | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
62 to 69.5 cm |