Tempshield™ Pink Cryo-Gloves™
Manufacturer:  Tempshield™ TSIPMAS
 View more versions of this product
Catalog No. 19-230-261
Specifications
Specifications
Fabrics, Multi-layered Insulated Construction
Fabric
Small, 8
Mid-Arm Length
13.5 to 15.25 in.
LNG (Liquefied Natural Gas), Liquid Helium, and Liquid Oxygen in enclosed systems, ultra-low temperature freezers, blast freezers, dry ice, and in coldrooms
Thermal
-259°F
Pink
34.5 to 39cm
Documents