Tempshield™ Pink Cryo-Gloves™
Manufacturer:  Tempshield™ TSIPEBXL
 View more versions of this product
Catalog No. 19-230-256
Specifications
Specifications
Fabrics, Multi-layered Insulated Construction
Fabric
X-Large, 11
Elbow Length
17.25 to 19.25 in.
LNG (Liquefied Natural Gas), Liquid Helium, and Liquid Oxygen in enclosed systems, ultra-low temperature freezers, blast freezers, dry ice, and in coldrooms
Thermal
-259°F
Pink
44 to 48.5cm
Documents