Tempshield™ Cryo-Gloves™
Breathable, stay-dry lining
Manufacturer:  Tempshield™ TSIMAMWP
 View more versions of this product
Catalog No. 11-394-326
Alert:
Not for immersion in liquid nitrogen or for use near open flame or in hazardous areas where ignition could occur.
Specifications Description & Specifications
Specifications
superior thermal protection when working with ultra low temperature freezers, blast freezers, dry ice and in coldrooms, also for use with Liquefied Natural Gas, liquid helium and liquid oxygen in enclosed systems
Waterproof Cryo-Gloves with Mid-Arm Length
-196°C
13.5 to 15.25 in.
Medium
-321°F
Blue
34.5 to 39cm