Please login to your online account to display your discounted pricing

Tempshield™ Cryo-Gloves™


Breathable, stay-dry lining

Manufacturer: tempshield™  TSISHL

 View more versions of this product

 Alert

Not for immersion in liquid nitrogen or for use near open flame or in hazardous areas where ignition could occur.

Catalog No. 11-394-308

  • / Pair
  • / Case of 10 Pair
 

Description & Specifications

Specifications

For Use With (Application) superior thermal protection when working with ultra low temperature freezers, blast freezers, dry ice and in coldrooms, also for use with Liquefied Natural Gas, liquid helium and liquid oxygen in enclosed systems
Length (English) 24.75 to 27 in.
Length (Metric) 62.5 to 68.5cm
Size Large
Style Cryo-Gloves with Shoulder Length
Temperature (English) Max. -321°F
Temperature (Metric) Max -196°C
Color Blue