Please login to your online account to display your discounted pricing

TEMED, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$166.38 - $243.62

Chemical Identifiers

Chemical Name or Material TEMED
CAS 110-18-9
Name Note Catalyst for Polyacrylamide Gels
Purity Grade Ultrapure Bioreagent
Molecular Formula (CH3)2NCH2CH2N(CH3)2
Formula Weight 116.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004455

 
avantor performance
409804
Narrow Mouth Amber Glass Bottle 100mL Each for $243.62

02004458

 
avantor performance
409802
Amber Glass Bottle 50mL Each for $166.38
Description & Specifications

Specifications

Formula Weight 116.21
Chemical Name or Material TEMED
Molecular Formula (CH3)2NCH2CH2N(CH3)2
Packaging Narrow Mouth Amber Glass Bottle
Quantity 100mL
CAS 110-18-9
Purity Grade Ultrapure Bioreagent
Flash Point 10.00°C
Specific Gravity 1L = 0.78kg
Name Note Catalyst for Polyacrylamide Gels