Please login to your online account to display your discounted pricing

TEMED, OmniPur™, Calbiochem™, EMD Millipore

Free radical initiator and stabilizer used in acrylamide polymerization for PAGE gels

$40.67 - $125.30

Chemical Identifiers

Chemical Name or Material TEMED
CAS 110-18-9
Physical Form Liquid
Molecular Formula C6H16N2
Formula Weight 116.2
Color Clear
Protease Not detected
RNase Not detected
DNase Not detected
Purity Grade Notes Molecular biology grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

892025ML

 
emd millipore corp
892025ML
Each for $40.67

8930100ML

 
emd millipore corp
8930100ML
Each for $125.30
Description & Specifications

Specifications

Formula Weight 116.2
Boiling Point 121°C
Melting Point −55°C
Quantity 25mL
Purity Grade Notes Molecular biology grade
Chemical Name or Material TEMED
DNase Not detected
Molecular Formula C6H16N2
CAS 110-18-9
UN Number UN 2372
Synonym N,N,N',N'-Tetramethylethylenediamine
Protease Not detected
RNase Not detected
Color Clear
Flash Point 20.85°C (69.5°F)
Odor Amine-like
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
DOT Information DOT Class: 3, Packaging Group: II
Physical Form Liquid
Solubility Information Water: Soluble
Safety and Handling

Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.