Please login to your online account to display your discounted pricing

TEMED (Electrophoresis), Fisher BioReagents


For polymerization of acrylamide/bis-acrylamide

Quantity 26mL (20g)
Packaging Amber Glass

Manufacturer: fisher scientific company  FLBP15020

 View more versions of this product

Catalog No. BP150-20

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

pH 8.0 to 8.5
Assay ≥97%
Melting Point −55°C
Boiling Point 120°C
Purity Grade Electrophoresis Grade
Formula Weight 116.21
Quantity 26mL (20g)
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless
Molecular Formula C6H16N2
Density 0.770g/cm³
DNase DNase free
Protease Protease free
Merck Index 15, 9277
UN Number 2372
Packaging Amber Glass
Purity Grade Notes DNase-, RNase- and Protease-Free
Linear Formula (CH3)2NCH2CH2N(CH3)2
ChemAlert Storage Symbol Red
Chemical Name or Material TEMED
Recommended Storage RT
Identification Pass Test
Electrophoresis Pass Test
Synonym N,N,N«,N«-Tetramethylethylenediamine
Assay Percent Range ≥97 % (by titration)
CAS 110-18-9

Specifications

Chemical Name or Material TEMED
CAS 110-18-9
Purity Grade Electrophoresis Grade
Purity Grade Notes DNase-, RNase- and Protease-Free
Physical Form Liquid
Assay ≥97%
Assay Percent Range ≥97 % (by titration)
Molecular Formula C6H16N2
Linear Formula (CH3)2NCH2CH2N(CH3)2
Formula Weight 116.21
Synonym N,N,N«,N«-Tetramethylethylenediamine
Accessories
 

RT

SDS & Safety Documents