Please login to your online account to display your discounted pricing

Tellurium Standard For ICP, 1000μg/mL in 5% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$182.31 - $331.06

Chemical Identifiers

Chemical Name or Material Tellurium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
View More Specs
Includes:  NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLTE2-2X

 
spex certiprep llc
PLTE22X
Triple-leached LDPE bottle 500mL Each for $331.06

PLTE2-2Y

 
spex certiprep llc
PLTE22Y
Triple-leached LDPE bottle 125mL Each for $182.31
Description & Specifications

Specifications

Concentration or Composition (by Analyte or Components) Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;1000μg/mL&tab;PLTE2-2Y, -2X
Quantity 500mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Health Hazard 1 May cause cancer. Causes skin irritation, serious eyeirritation. Depleted radioactive material.
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Solvent or Matrix 5% HNO3
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Chemical Name or Material Tellurium Standard
CAS Max % 0.1
CAS 7697-37-2
CAS Min % 5.0
CAS Min % 94.9
CAS Max % 94.9
CAS 13494-80-9
CAS Min % 0.1
CAS 7732-18-5
CAS Max % 5.0

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Te1000μg/mL

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Safety and Handling

May cause cancer. Causes skin irritation, serious eyeirritation. Depleted radioactive material.

Ambient