Please login to your online account to display your discounted pricing

Tellurium Standard For ICP, 1000μg/mL in 5% HNO3, Assurance Grade, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 500mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLTE22X

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLTE2-2X

  • / Each
 

Description & Specifications

Chemical Identifiers

Concentration or Composition (by Analyte or Components) Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;1000μg/mL&tab;PLTE2-2Y, -2X
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Packaging Triple-leached LDPE bottle
DOT Information Hazardous
Solvent or Matrix 5% HNO3
Quantity 500mL
Health Hazard 1 May cause cancer. Causes skin irritation, serious eyeirritation. Depleted radioactive material.
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Chemical Name or Material Tellurium Standard
CAS 7697-37-2
CAS Min % 5.0
CAS Max % 0.1
CAS Min % 94.9
CAS Max % 94.9
CAS Min % 0.1
CAS 13494-80-9
CAS 7732-18-5
CAS Max % 5.0

Specifications

Chemical Name or Material Tellurium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Te1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

May cause cancer. Causes skin irritation, serious eyeirritation. Depleted radioactive material.

Ambient