Please login to your online account to display your discounted pricing

Tellurium Standard, 1000mg/L in 5% HNOHNO3, (Assurance Grade), SPEX CertiPrep™


Quantity 30mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  SPXPLTE22M

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Catalog No. PLTE22M

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 30mL
Chemical Name or Material Tellurium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
DOT Information Nonhazardous
Solvent or Matrix 5% HNO3
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.

Specifications

Chemical Name or Material Tellurium Standard

AnalyteConcentration
Te1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Ambient