missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium Powder, -200 mesh, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1107530

 View more versions of this product

Catalog No. AA1107530



Specifications

Specifications

Tellurium
99.999% (Metals basis)
MFCD00134062
and compounds
[Te]
127.6
CHEBI:30452
Odorless
250g
13494-80-9
Te
Gray powder
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium
6327182
Powder
Gray

Chemical Identifiers

13494-80-9
127.6
PORWMNRCUJJQNO-UHFFFAOYSA-N
6327182
tellurium
Te
MFCD00134062
and compounds
CHEBI:30452
[Te]
Safety and Handling

Safety and Handling

H301-H332-H315-H319-H335; Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 236-813-4

Documents