missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tellurium powder, -200 mesh, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 1060518

 View more versions of this product

Catalog No. AA1060518



Specifications

Specifications

Tellurium powder
Gray
13494-80-9
(metals basis)
MFCD00134062
and compounds
[Te]
127.6
CHEBI:30452
99.5%
−200 Mesh
50g
Odorless
Te
UN3288
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium
6327182
Powder

Chemical Identifiers

13494-80-9
127.6
PORWMNRCUJJQNO-UHFFFAOYSA-N
6327182
tellurium
Te
MFCD00134062
and compounds
CHEBI:30452
[Te]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : WY2625000

TSCA : TSCA

Documents