missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium, plasma standard solution, Specpure™, Te 1000μg/ml, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 44632AD

 View more versions of this product

Catalog No. AA44632AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Tellurium, plasma standard solution, Specpure™
MFCD00134062
Te 1000μg/ml
50mL
Te in 5% HNO3
UN3264
Liquid

Chemical Identifiers

Te in 5% HNO3
MFCD00134062
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318;
Causes severe skin burns and eye damage.
Causes serious eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents