missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tellurium, plasma standard solution, Specpure™, Te 10,000μg/mL

Manufacturer:  Alfa Aesar™ 47187AD

 View more versions of this product

Catalog No. AA47187AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Tellurium, plasma standard solution
Te in 30% HCl
UN1789
Specpure™
50mL
Te 10,000μg/mL
MFCD00134062
Liquid
Acidic

Chemical Identifiers

MFCD00134062
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: HYDROCHLORIC ACID

TSCA : TSCA

SDS Documents