missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tellurium lump, typically 12mm (0.47 in.), 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 1075809

 View more versions of this product

Catalog No. AA1075809



Specifications

Specifications

Tellurium lump
Typically 12mm (0.47 in.)
Odorless
Te
and compounds
[Te]
127.6
CHEBI:30452
99.9999%
10g
13494-80-9
(metals basis)
MFCD00134062
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium
6327182
Lumps

Chemical Identifiers

13494-80-9
127.6
PORWMNRCUJJQNO-UHFFFAOYSA-N
6327182
tellurium
Te
MFCD00134062
and compounds
CHEBI:30452
[Te]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents