missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium lump, typically 12mm (0.47 in.), 99.9999% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01075818

 View more versions of this product

Catalog No. AA1075818



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tellurium lump
13494-80-9
Te
50 g
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium
6327182
99.9999%
Lump
Typically 12mm (0.47 in.)
(metals basis)
MFCD00134062
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
[Te]
127.60
CHEBI:30452
Odorless

Chemical Identifiers

13494-80-9
127.60
PORWMNRCUJJQNO-UHFFFAOYSA-N
6327182
tellurium
Te
MFCD00134062
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
CHEBI:30452
[Te]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

P264b-P270-P301+P310-P330-P501c

H301

EINECSNumber : 236-813-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only